Pyrrolo[1,2-a]pyrazine 97%

Supplier: Ambeed
Warning

A521118-1G A521118-5G A521118-250MG
103828-508EA 245 USD
103828-508 103839-616 76843-616 76843-618 77034-666
Pyrrolo[1,2-a]pyrazine 97%
Pyrrolo[1,2-a]pyrazine

Precaución: for Research Use Only

Formula: C7H6N2
MW: 118.1400 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01605573
CAS Number: 274-45-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR