Pyrrolidin-2-ylmethanol 98%

Supplier: Ambeed
Warning

Synonyms: DL-Prolinol, 2-Pyrrolidinemethanol, 2-(Hydroxymethyl)pyrrolidine

A175886-100G A175886-10G A175886-25G A175886-5G A175886-1G
103807-096EA 5 USD
103807-096 103845-408 76784-440 76784-442 77082-756 77082-758 77301-406
Pyrrolidin-2-ylmethanol 98%
Pyrrolidin-2-ylmethanol

Precaución: for Research Use Only

Formula: C5H11NO
MW: 101.15 g/mol
Boiling Pt: 211 °C (760 mm Hg)
Flash Pt: 86.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00601073
CAS Number: 498-63-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR