Pyrimidine-5-carboxylic acid 97%

Supplier: Ambeed
Warning

A185811-100G A185811-10G A185811-250MG A185811-1G A185811-25G A185811-5G
103815-888EA 10 USD
103815-888 103846-458 77012-102 77012-104 77012-106 77084-092 77084-094 77084-096
Pyrimidine-5-carboxylic acid 97%
Pyrimidine-5-carboxylic acid

Precaución: for Research Use Only

Formula: C5H4N2O2
Boiling Pt: 318.7 °C (760 mm Hg)
Melting Pt: 259 °C
Flash Pt: 146.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00856162
CAS Number: 4595-61-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR