Pyrimidine-4-carboxylic acid 97%

Supplier: Ambeed
Warning

A193086-100G A193086-10G A193086-500G A193086-1G A193086-5G A193086-25G
103816-496EA 9 USD
103816-496 103847-112 76999-044 76999-046 76999-048 77085-008 77085-010 77302-324
Pyrimidine-4-carboxylic acid 97%
Pyrimidine-4-carboxylic acid

Precaución: for Research Use Only

Formula: C5H4N2O2
MW: 124.10 g/mol
Boiling Pt: 318.7 °C (760 mm Hg)
Melting Pt: 229.8…231.9 °C
Flash Pt: 146.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00129737
CAS Number: 31462-59-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR