Pyrimidine-4,6-dicarboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: Pyrimidine-4,6-dicarboxylic acid

A133911-100G A133911-10G A133911-1G A133911-25G A133911-5G
103801-930EA 7 USD
103801-930 76972-248 76972-250 77314-626 77314-628 77314-630
Pyrimidine-4,6-dicarboxylic acid 95%
Pyrimidine-4,6-dicarboxylic acid

Precaución: for Research Use Only

Formula: C₆H₄N₂O₄
MW: 168.107 g/mol
Boiling Pt: 497.4 °C (760 mm Hg)
Melting Pt: 238…240 °C
Flash Pt: 254.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00094473
CAS Number: 16490-02-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR