Pyrimidine-4,5,6-triamine 97%

Supplier: Ambeed
Warning

A151972-10G A151972-250MG A151972-1G A151972-5G
103804-330EA 44 USD
103804-330 103842-480 77110-346 77110-348 77318-074 77318-076
Pyrimidine-4,5,6-triamine 97%
Pyrimidine-4,5,6-triamine

Precaución: for Research Use Only

Formula: C₄H₇N₅
Boiling Pt: 406.2 °C (760 mm Hg)
Flash Pt: 228.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00023271
CAS Number: 118-70-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR