5-Pyrimidineboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: Pyrimidine-5-boronic acid, 5-Pyrimidylboronic acid, Pyrimidin-5-ylboronic acid, 5-Pyrimidinylboronic acid

A390529-1G A390529-10G A390529-25G A390529-5G A390529-100G A390529-500G
103826-866EA 9 USD
103826-866 103837-908 76841-298 76876-772 77306-998 77307-000 77672-944 77672-946
5-Pyrimidineboronic acid 98%
5-Pyrimidineboronic acid

Precaución: for Research Use Only

Formula: C₄H₅BN₂O₂
Boiling Pt: 334.7 °C (760 mm Hg)
Melting Pt: 110…114 °C
Flash Pt: 156.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03002366
CAS Number: 109299-78-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR