4(3H)-Pyrimidinone 97%

Supplier: Ambeed
Warning

Synonyms: 4-Pyrimidinol, 4(3H)-Pyrimidone, 3H-Pyrimidin-4-one, 4-Hydroxypyrimidine, Pyrimidin-4(3H)-one

A202249-100G A202249-10G A202249-1G A202249-25G A202249-5G
103817-366EA 6 USD
103817-366 103848-002 76975-278 76975-280 77086-100 77086-102 77086-104
4(3H)-Pyrimidinone 97%
4(3H)-Pyrimidinone

Precaución: for Research Use Only

Formula: C₄H₄N₂O
MW: 96.09 g/mol
Boiling Pt: 184.6 °C (760 mm Hg)
Melting Pt: 163…167 °C
Flash Pt: 65.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00006664
CAS Number: 4562-27-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR