2-Hydroxypyrimidine hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: Pyrimidin-2-ol hydrochloride, 2-Pyrimidinol Hydrochloride, 2(1H)-pyrimidinone monohydrochloride

A115847-10G A115847-25G A115847-500G
103824-494EA 5 USD
103824-494 76971-154 76971-156 76971-158
2-Hydroxypyrimidine hydrochloride 98%
2-Hydroxypyrimidine hydrochloride

Precaución: for Research Use Only

Formula: C₄H₄N₂O
MW: 132.55 g/mol
Boiling Pt: 221.2 °C (760 mm Hg)
Melting Pt: 200…205 °C
Flash Pt: 87.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00012781
CAS Number: 38353-09-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR