Pyrido[3,4-d]pyrimidin-4(3H)-one 95%

Supplier: Ambeed
Warning

A179115-100MG A179115-250MG A179115-1G
103807-396EA 70 USD
103807-396 103845-738 76784-524 76784-526 77301-594
Pyrido[3,4-d]pyrimidin-4(3H)-one 95%
Pyrido[3,4-d]pyrimidin-4(3H)-one

Precaución: for Research Use Only

Formula: C7H5N3O
MW: 147.1300 g/mol
Melting Pt: 305 °C
Storage Temperature: Ambient
MDL Number: MFCD09999183
CAS Number: 19178-25-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR