Pyridino[3,2-b]pyrazine 98%

Supplier: Ambeed
Warning

A508902-1G A508902-5G A508902-100MG A508902-250MG
103828-360EA 61 USD
103828-360 103839-460 76843-448 76843-450 77144-080 77144-082
Pyridino[3,2-b]pyrazine 98%
Pyridino[3,2-b]pyrazine

Precaución: for Research Use Only

Formula: C₇H₅N₃
Boiling Pt: 253.9 °C (760 mm Hg)
Melting Pt: 139…143 °C
Storage Temperature: Ambient
MDL Number: MFCD00006717
CAS Number: 322-46-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR