3-Pyridinesulfonic acid 97%

Supplier: Ambeed
Danger

Synonyms: Pyridine-3-sulphonic acid

A494914-25G A494914-5G A494914-100G
103810-390EA 293 USD
103810-390 103839-324 76843-290 77307-592 77673-290
3-Pyridinesulfonic acid 97%
3-Pyridinesulphonic acid

Precaución: for Research Use Only

Formula: C₅H₅NO₃S
Melting Pt: >300 °C
Flash Pt: 66 °C
Storage Temperature: Ambient
MDL Number: MFCD00006381
CAS Number: 636-73-7
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR