Pyridine-3-sulfonamide 98%

Supplier: Ambeed
Warning

Synonyms: 3-Sulphamoylpyridine, 3-(Aminosulphonyl)pyridine

A527014-5G A527014-100MG A527014-250MG
103828-542EA 43 USD
103828-542 103839-654 76843-664 77292-296 77292-298
Pyridine-3-sulfonamide 98%
Pyridine-3-sulfonamide

Precaución: for Research Use Only

Formula: C₅H₆N₂O₂S
MW: 158.178 g/mol
Boiling Pt: 357.7±34.0 °C (760 mm Hg)
Flash Pt: 170.1 °C
Storage Temperature: Ambient
MDL Number: MFCD08690712
CAS Number: 2922-45-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR