Pyridine-3,5-dicarboxylic acid, (max. 5% H₂O) 98% (contains < 5% H₂O)
Supplier: Ambeed
|
Warning
|
A280305-25G
A280305-100G
A280305-10G
A280305-1000G
A280305-500G
103853-450EA
7
USD
103853-450
77093-384
77141-146
77141-148
77685-790
77685-791
Pyridine-3,5-dicarboxylic acid, (max. 5% H₂O) 98% (contains < 5% H₂O)
Pyridine-3,5-dicarboxylic acid
Precaución: for Research Use Only
Formula:
C₇H₅NO₄ Boiling Pt: 463.7 °C (760 mm Hg) Melting Pt: >300 °C Flash Pt: 234.2 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006393 CAS Number: 499-81-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...