3,4-Pyridinedimethanol 97%

Supplier: Ambeed
Warning

Synonyms: Pyridine-3,4-diyldimethanol

A149950-250MG A149950-1G A149950-5G
103804-110EA 35 USD
103804-110 103842-238 77150-352 77299-766 77299-768
3,4-Pyridinedimethanol 97%
3,4-Pyridinedimethanol

Precaución: for Research Use Only

Formula: C₇H₉NO₂
MW: 139.16 g/mol
Boiling Pt: 344.4 °C (760 mm Hg)
Melting Pt: 144 °C
Flash Pt: 162.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00160202
CAS Number: 38070-80-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR