3,4-Diaminopyridine 98%

Supplier: Ambeed
Danger

A101083-100MG A101083-1G A101083-10G A101083-5G
103795-796EA 9 USD
103795-796 103821-486 77282-656 77296-982 77614-740 77614-742
3,4-Diaminopyridine 98%
3,4-Diaminopyridine

Precaución: for Research Use Only

Formula: C₅H₇N₃
Boiling Pt: 369.3 °C (760 mm Hg)
Melting Pt: 216…218 °C
Flash Pt: 240 °C
Storage Temperature: Ambient
MDL Number: MFCD00006401
CAS Number: 54-96-6
UN: 2811
ADR: 6.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR