Pyridine-2-sulfonyl Fluoride 97%

Supplier: Ambeed
Danger

Synonyms: PyFluor

A650302-10G A650302-250MG A650302-1G A650302-25G A650302-5G
103840-396EA 41 USD
103840-396 76727-324 76727-326 76844-558 76844-560 76844-562
Pyridine-2-sulfonyl Fluoride 97%
Pyridine-2-sulphonyl Fluoride

Precaución: for Research Use Only

Formula: C₅H₄FNO₂S
MW: 161.15 g/mol
Boiling Pt: 234.9 °C (760 mm Hg)
Flash Pt: 95.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09265513
CAS Number: 878376-35-3
UN: 3265
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR