Pyridine-2,6-dicarbonitrile 97%

Supplier: Ambeed
Warning

Synonyms: 2,6-Dicyanopyridine

A257648-1G A257648-25G A257648-5G A257648-100G A257648-10G A257648-250MG A257648-500G
103812-326EA 8 USD
103812-326 103852-194 77091-706 77091-708 77091-710 77250-082 77250-084 77250-086 77250-088
Pyridine-2,6-dicarbonitrile 97%
Pyridine-2,6-dicarbonitrile

Precaución: for Research Use Only

Formula: C₇H₃N₃
MW: 129.12 g/mol
Boiling Pt: 334.9 °C (760 mm Hg)
Melting Pt: 137…139 °C
Flash Pt: 107.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00129021
CAS Number: 2893-33-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR