Pyridine-2,4-dicarboxylic acid 99%

Supplier: Ambeed
Warning

A144466-10G A144466-500G A144466-1G A144466-5G A144466-100G
103803-410EA 6 USD
103803-410 103841-486 77272-632 77272-634 77316-732 77316-734 77615-158
Pyridine-2,4-dicarboxylic acid 99%
Pyridine-2,4-dicarboxylic acid

Precaución: for Research Use Only

Formula: C₇H₅NO₄
Boiling Pt: 574.8 °C (760 mm Hg)
Melting Pt: 243…246 °C
Flash Pt: 301.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00149413
CAS Number: 499-80-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR