Pyridine-2,4-diamine 97%

Supplier: Ambeed
Warning

A202971-1G A202971-250MG
103817-432EA 47 USD
103817-432 103848-080 77302-840 77667-692
Pyridine-2,4-diamine 97%
Pyridine-2,4-diamine

Precaución: for Research Use Only

Formula: C5H7N3
Boiling Pt: 358 °C (760 mm Hg)
Flash Pt: 197.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00234953
CAS Number: 461-88-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR