4-(Aminomethyl)pyridine 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
4-Picolamine, (4-Pyridyl)methylamine, Pyridin-4-ylmethanamine, 4-Picolylamine, 4-Pyridylmethylamine
A370655-25G
A370655-10G
A370655-5G
A370655-100G
103837-476EA
8
USD
103837-476
76840-724
77101-568
77306-812
77613-100
4-(Aminomethyl)pyridine 98%
4-(Aminomethyl)pyridine
Precaución: for Research Use Only
Formula:
C₆H₈N₂ MW: 108.14 g/mol Boiling Pt: 230 °C (760 mm Hg) Melting Pt: …8 °C Flash Pt: 104 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006449 CAS Number: 3731-53-1 UN: 2735 ADR: 8,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...