4-Pyridineboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-Pyridinylboronic acid, Pyridine-4-boronic acid, Pyridin-4-ylboronic acid

A106578-10G A106578-25G A106578-1G A106578-5G A106578-100G
103797-044EA 6 USD
103797-044 103822-798 77270-590 77270-592 77309-656 77309-658 77570-390
4-Pyridineboronic acid 98%
4-Pyridineboronic acid

Precaución: for Research Use Only

Formula: C₅H₆BNO₂
MW: 122.92 g/mol (anhy)
Melting Pt: >300 °C
Flash Pt: 153.8 °C
Storage Temperature: Freezer
MDL Number: MFCD01074545
CAS Number: 1692-15-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR