3-(Aminomethyl)pyridine 97%

Supplier: Ambeed
Danger

Synonyms: 3-Pyridylmethylamine, (3-Pyridyl)methylamine, 3-Picolamine, Pyridin-3-ylmethanamine, 3-Picolylamine

A310850-25G A310850-10G A310850-500G
103810-070EA 241 USD
103810-070 103835-826 76838-600 76977-898 76977-900
3-(Aminomethyl)pyridine 97%
3-(Aminomethyl)pyridine

Precaución: For research use only

Formula: C₆H₈N₂
Boiling Pt: 226 °C (760 mm Hg)
Melting Pt: …21 °C
Flash Pt: 108 °C
Storage Temperature: Ambient
MDL Number: MFCD00006412
CAS Number: 3731-52-0
UN: 2735
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR