3-Pyridineboronic acid 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Pyridinylboronic acid, Pyridin-3-ylboronic acid, Pyridine-3-boronic acid
A106751-10G
A106751-25G
A106751-500G
A106751-1G
A106751-5G
A106751-100G
103797-070EA
6
USD
103797-070
103822-828
77008-640
77008-642
77008-644
77283-510
77283-512
77549-048
3-Pyridineboronic acid 95%
3-Pyridineboronic acid
Precaución: for Research Use Only
Formula:
C₅H₆BNO₂ Melting Pt: >300 °C Flash Pt: 140.5 °C Storage Temperature: Ambient |
MDL Number:
MFCD00674177 CAS Number: 1692-25-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...