3-Pyridone 98%

Supplier: Ambeed
Warning

Synonyms: 3-Hydroxypyridine, β-Pyridone, Pyridin-3-ol, 3-Pyridinol

A141983-100G A141983-10G A141983-25G A141983-500G A141983-5G A141983-1000G
103808-622EA 69 USD
103808-622 77010-372 77010-374 77010-376 77010-378 77010-380 77299-264
3-Pyridone 98%
3-Pyridone

Precaución: for Research Use Only

Formula: C₅H₅NO
MW: 95.1 g/mol
Boiling Pt: 151…153 °C (760 mm Hg)
Melting Pt: 125…128 °C
Flash Pt: 173 °C
Storage Temperature: Ambient
MDL Number: MFCD00006378
CAS Number: 109-00-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR