Pyridazine-4,5-dicarboxylic acid 98%

Supplier: Ambeed
Warning

A260260-5G A260260-1G A260260-25G
103812-488EA 117 USD
103812-488 103852-370 77091-956 77304-782 77883-717
Pyridazine-4,5-dicarboxylic acid 98%
Pyridazine-4,5-dicarboxylic acid

Precaución: for Research Use Only

Formula: C₆H₄N₂O₄
Boiling Pt: 539.4 °C (760 mm Hg)
Melting Pt: >210 °C
Flash Pt: 280 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06742784
CAS Number: 59648-14-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR