Pyridazine-4,5-diamine 95%

Supplier: Ambeed
Warning

A120583-100MG A120583-250MG A120583-50MG
103799-962EA 688 USD
103799-962 77026-388 77026-390 77026-392
Pyridazine-4,5-diamine 95%
Pyridazine-4,5-diamine
Formula: C₄H₆N₄
Boiling Pt: 412.5 °C (760 mm Hg)
Melting Pt: 260…264 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11110431
CAS Number: 28682-70-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR