Pyridazin-4-amine 97%

Supplier: Ambeed
Warning

A224092-100G A224092-10G A224092-250MG A224092-1G A224092-25G A224092-5G
103819-082EA 18 USD
103819-082 103849-972 77013-342 77013-344 77013-346 77088-606 77303-684 77303-686
Pyridazin-4-amine 97%
Pyridazin-4-amine

Precaución: for Research Use Only

Formula: C4H5N3
Boiling Pt: 326.2 °C (760 mm Hg)
Flash Pt: 177.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00233975
CAS Number: 20744-39-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR