3(2H)-Pyridazinone 97%

Supplier: Ambeed
Warning

Synonyms: 2,3-dihydropyridazin-3-one, Pyridazin-3(2H)-one

A135054-100G A135054-10G A135054-500G A135054-1G A135054-25G A135054-5G
103802-084EA 6 USD
103802-084 77109-468 77109-470 77109-472 77298-750 77314-864 77314-866
3(2H)-Pyridazinone 97%
3(2H)-Pyridazinone

Precaución: for Research Use Only

Formula: C₄H₄N₂O
MW: 96.0874 g/mol
Boiling Pt: 315.9 °C (760 mm Hg)
Melting Pt: 98…104 °C
Flash Pt: 144.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00099605
CAS Number: 504-30-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR