1-Aminopyrene 97%

Supplier: Ambeed
Warning

Synonyms: Pyren-1-ylamine, Pyren-1-amine

A321565-1G A321565-25G A321565-5G A321565-100G A321565-250MG
103825-280EA 25 USD
103825-280 103836-214 76839-120 76839-122 76839-124 77032-778 77032-780
1-Aminopyrene 97%
1-Aminopyrene

Precaución: for Research Use Only

Formula: C₁₆H₁₁N
MW: 217.27 g/mol
Boiling Pt: 440.8 °C (760 mm Hg)
Melting Pt: 114…118 °C at N/A mmHg
Flash Pt: 246.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00004140
CAS Number: 1606-67-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR