Pyrazolo[1,5-a]pyrimidine-3-carboxylic acid 97%

Supplier: Ambeed
Warning

A279132-1G A279132-25G A279132-5G A279132-100G A279132-10G A279132-250MG
103813-454EA 7 USD
103813-454 103853-398 77093-326 77093-328 77093-330 77166-612 77166-614 77166-616
Pyrazolo[1,5-a]pyrimidine-3-carboxylic acid 97%
Pyrazolo[1,5-a]pyrimidine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₅N₃O₂
Melting Pt: 180 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01975194
CAS Number: 25940-35-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR