Pyrazolo[1,5-a]pyrimidin-5-ol 98%
Supplier: Ambeed
|
Warning
|
A103418-100MG
A103418-1G
A103418-5G
A103418-25G
A103418-250MG
103796-548EA
8
USD
103796-548
103822-254
76780-638
77309-122
77309-124
77871-394
77871-393
Pyrazolo[1,5-a]pyrimidin-5-ol 98%
Precaución: for Research Use Only
Formula:
C₆H₅N₃O Storage Temperature: Ambient |
MDL Number:
MFCD04035685 CAS Number: 29274-22-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...