Pyrazolo[1,5-a]pyridine-3-carboxylic acid 95%

Supplier: Ambeed
Warning

A236058-250MG A236058-1G A236058-5G A236058-100MG
103811-064EA 19 USD
103811-064 103850-834 77013-652 77089-868 77089-870 77683-698
Pyrazolo[1,5-a]pyridine-3-carboxylic acid 95%
Pyrazolo[1,5-a]pyridine-3-carboxylic acid

Precaución: For research use only.

Formula: C₈H₆N₂O₂
MW: 162.1454 g/mol
Melting Pt: 199…201 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00229214
CAS Number: 16205-46-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR