Pyrazolo[1,5-a]pyridine-2-carboxylic acid 98%

Supplier: Ambeed
Warning

A101463-10G A101463-250MG A101463-5G
103795-984EA 26 USD
103795-984 103821-668 76809-244 76809-246 77297-110
Pyrazolo[1,5-a]pyridine-2-carboxylic acid 98%
Pyrazolo[1,5-a]pyridine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₆N₂O₂
MW: 162.1500 g/mol
Melting Pt: 189…192 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06739057
CAS Number: 63237-88-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR