1H-Pyrazole-3-carbaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: 3-Formylpyrazole, Pyrazole-3-carboxaldehyde, 3-Formyl-1H-pyrazole

A115602-100G A115602-1G A115602-10G A115602-25G
103798-594EA 11 USD
103798-594 103824-452 77160-990 77311-270 77549-290 77566-076
1H-Pyrazole-3-carbaldehyde 97%
1H-Pyrazole-3-carbaldehyde

Precaución: For research use only.

Formula: C₄H₄N₂O
MW: 96.09 g/mol
Boiling Pt: 299.9 °C (760 mm Hg)
Melting Pt: 142…144 °C
Flash Pt: 139.672 °C
Storage Temperature: Ambient
MDL Number: MFCD00129925
CAS Number: 3920-50-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR