Pyrazine-2-carbaldehyde 95%
Supplier: Ambeed
|
Warning
|
A199037-1G
A199037-25G
A199037-5G
A199037-100MG
A199037-10G
A199037-250MG
103817-072EA
112
USD
103817-072
103847-696
77085-730
77085-732
77085-734
77164-520
77164-522
77164-524
Pyrazine-2-carbaldehyde 95%
Pyrazine-2-carbaldehyde
Precaución: for Research Use Only
Formula:
C5H4N2O MW: 108.0981 g/mol Flash Pt: 70.5 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD02179278 CAS Number: 5780-66-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...