Pyrazine-2,6-diamine hydrochloride 95%

Supplier: Ambeed
Warning

A233614-10G A233614-25G A233614-1G A233614-5G
103810-892EA 46 USD
103810-892 103850-654 76815-232 76815-234 77089-614 77089-616
Pyrazine-2,6-diamine hydrochloride 95%
Pyrazine-2,6-diamine hydrochloride

Precaución: for Research Use Only

Formula: C₄H₇ClN₄
Storage Temperature: Refrigerator
MDL Number: MFCD21604008
CAS Number: 1370411-48-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR