Pyrazine-2,5-dicarboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2,5-Pyrazinedicarboxylic acid

A344554-100G A344554-10G A344554-1G A344554-25G A344554-5G
103825-908EA 7 USD
103825-908 103836-888 76724-382 76724-384 76839-936 76839-938 76839-940
Pyrazine-2,5-dicarboxylic acid 97%
Pyrazine-2,5-dicarboxylic acid

Precaución: for Research Use Only

Formula: C₆H₄N₂O₄
MW: 168.108 g/mol
Boiling Pt: 466.5 °C (760 mm Hg)
Melting Pt: 255…260 °C
Flash Pt: 236 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01630902
CAS Number: 122-05-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR