Pyrazine-2,3-diol 95%

Supplier: Ambeed
Warning

A256481-100MG A256481-250MG A256481-1G A256481-5G
103812-268EA 240 USD
103812-268 103852-132 76722-680 76722-682 77879-473 77879-474
Pyrazine-2,3-diol 95%
Pyrazine-2,3-diol

Precaución: for Research Use Only

Formula: C₄H₄N₂O₂
Storage Temperature: Freezer
MDL Number: MFCD00085020
CAS Number: 221303-71-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR