Pyrazin-2-ylmethanol 98%
Supplier: Ambeed
|
Warning
|
A501507-1G
A501507-5G
A501507-10G
A501507-25G
103828-296EA
13
USD
103828-296
103839-384
76843-350
76843-352
77279-688
77279-690
Pyrazin-2-ylmethanol 98%
Pyrazin-2-ylmethanol
Precaución: for Research Use Only
Formula:
C5H6N2O Boiling Pt: 231.9 °C (760 mm Hg) Melting Pt: 38.5 °C Flash Pt: 94.1 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD01693938 CAS Number: 6705-33-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...