Pyrazin-2-ylboronic acid 95%

Supplier: Ambeed
Warning

A202067-100MG A202067-250MG A202067-5G
103792-378EA 173 USD
103792-378 103817-354 76975-272 76975-274 76975-276
Pyrazin-2-ylboronic acid 95%
Pyrazin-2-ylboronic acid

Precaución: for Research Use Only

Formula: C₄H₅BN₂O₂
MW: 123.91 g/mol
Boiling Pt: 329.2±45.0 °C (760 mm Hg)
Flash Pt: 152.9 °C
Storage Temperature: Freezer
MDL Number: MFCD08703263
CAS Number: 762263-64-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR