2-Aminopyrazine 98%

Supplier: Ambeed
Warning

Synonyms: Aminopyrazine, Pyrazin-2-amine

A186374-100G A186374-10G A186374-500G A186374-1000G A186374-25G
103809-202EA 227 USD
103809-202 103846-516 76872-916 76872-918 76872-920 77084-176 77084-178
2-Aminopyrazine 98%
2-Aminopyrazine

Precaución: for Research Use Only

Formula: C₄H₅N₃
MW: 95.1 g/mol
Boiling Pt: 250.905 °C (760 mm Hg)
Melting Pt: 117…120 °C
Flash Pt: 129.08 °C
Storage Temperature: Ambient
MDL Number: MFCD00006137
CAS Number: 5049-61-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR