7-Hydroxy-3-(4-hydroxyphenyl)-8-((2S,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)-4H-chromen-4-one 97%

Supplier: Ambeed
Warning

A228947-100G A228947-10G A228947-1G A228947-25G A228947-5G
103819-430EA 6 USD
103819-430 103850-334 77288-328 77288-330 77303-836 77303-838 77303-840
7-Hydroxy-3-(4-hydroxyphenyl)-8-((2S,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)-4H-chromen-4-one 97%
7-Hydroxy-3-(4-hydroxyphenyl)-8-((2S,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)-4H-chromen-4-one

Precaución: for Research Use Only

Formula: C₂₁H₂₀O₁₀
MW: 432.38 g/mol
Boiling Pt: 791.2 °C (760 mm Hg)
Melting Pt: 187…189 °C
Flash Pt: 281.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00063399
CAS Number: 3681-99-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR