(4-Methylphenyl)methanesulfonyl chloride 96%

Supplier: Ambeed
Danger

Synonyms: p-Tolylmethanesulphonyl chloride, α-p-Xylenesulphonyl chloride, 4-Methylbenzylsulphonyl chloride

A839008-10G A839008-250MG A839008-5G
103841-200EA 46 USD
103841-200 76728-776 76728-778 76845-438
(4-Methylphenyl)methanesulfonyl chloride 96%
(4-Methylphenyl)methanesulphonyl chloride

Precaución: for Research Use Only

Formula: C₈H₉ClO₂S
MW: 204.67 g/mol
Boiling Pt: 305.2 °C (760 mm Hg)
Melting Pt: 80…85 °C
Flash Pt: 138.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01631931
CAS Number: 51419-59-1
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR