PT-2385 98%

Supplier: Ambeed
Warning

Synonyms: PT2385, (S)-3-((2,2-difluoro-1-hydroxy-7-(methylsulphonyl)-2,3-dihydro-1H-inden-4-yl)oxy)-5-fluorobenzonitrile

A468981-50MG A468981-10MG A468981-1MG A468981-25MG A468981-5MG
103791-678EA 146 USD
103791-678 103795-158 103799-654 76842-984 77241-212 77241-214 77241-216 77241-218
PT-2385 98%
PT-2385

Precaución: for Research Use Only

Formula: C₁₇H₁₂F₃NO₄S
MW: 383.34 g/mol
Storage Temperature: Freezer
MDL Number: MFCD28963916
CAS Number: 1672665-49-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR