3,4-Dihydroxybenzoic acid 98%

Supplier: Ambeed
Warning

Synonyms: Protocatechuic acid

A202277-25G A202277-5G A202277-100G A202277-500G A202277-1000G A202277-1G
103848-006EA 6 USD
103848-006 77086-110 77086-112 77139-364 77139-366 77550-298 77567-928
3,4-Dihydroxybenzoic acid 98%
3,4-Dihydroxybenzoic acid

Precaución: For research use only.

Formula: C₇H₆O₄
MW: 154.12 g/mol
Melting Pt: 197…200 °C
Storage Temperature: Ambient
MDL Number: MFCD00002509
CAS Number: 99-50-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR