6-Propyl-2-thiouracil 97%

Supplier: Ambeed
Warning

Synonyms: Propylthiouracil, 4-Hydroxy-2-mercapto-6-propylpyrimidine, 4-Propyl-2-thiouracil, 6-Propyl-2-thioxo-2,3-dihydropyrimidin-4(1H)-one

A115152-1000G A115152-100G A115152-100MG A115152-25G A115152-500G
103824-368EA 5 USD
103824-368 77108-214 77108-216 77108-218 77108-220 77108-222
6-Propyl-2-thiouracil 97%
6-Propyl-2-thiouracil

Precaución: for Research Use Only

Formula: C₇H₁₀N₂OS
MW: 170.24 g/mol
Melting Pt: 218…220 °C
Storage Temperature: Ambient
MDL Number: MFCD00006041
CAS Number: 51-52-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR