1-Propylboronic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Propylboronic acid
A224397-100G
A224397-10G
A224397-25G
A224397-5G
A224397-1G
103819-106EA
6
USD
103819-106
103849-996
76975-936
76975-938
77088-648
77088-650
77303-696
1-Propylboronic acid 98%
1-Propylboronic acid
Precaución: for Research Use Only
Formula:
C₃H₉BO₂ Boiling Pt: 170.8 °C (760 mm Hg) Melting Pt: 105…108 °C Storage Temperature: Freezer |
MDL Number:
MFCD01074564 CAS Number: 17745-45-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...