Isopropylsulfonamide 98%

Supplier: Ambeed
Warning

Synonyms: Propane-2-sulphonamide, 2-Sulphamoylpropane

A405585-25G A405585-100MG A405585-250MG A405585-10G
103827-120EA 12 USD
103827-120 103838-160 76841-648 76876-936 76876-938 77672-980
Isopropylsulfonamide 98%
Isopropylsulfonamide

Precaución: for Research Use Only

Formula: C₃H₉NO₂S
MW: 123.174 g/mol
Boiling Pt: 216.5 °C (760 mm Hg)
Flash Pt: 84.7 °C
Storage Temperature: Ambient
MDL Number: MFCD03550610
CAS Number: 81363-76-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR