Propane-1,3-diylbis(dicyclohexylphosphonium) tetrafluoroborate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
dcpp
A200643-100MG
A200643-10G
A200643-250MG
A200643-1G
A200643-25G
A200643-5G
103817-228EA
9
USD
103817-228
103847-860
76986-434
76986-436
76986-438
77085-920
77085-922
77085-924
Propane-1,3-diylbis(dicyclohexylphosphonium) tetrafluoroborate 97%
Propane-1,3-diylbis(dicyclohexylphosphonium) tetrafluoroborate
Precaución: for Research Use Only
Formula:
C₂₇H₅₂B₂F₈P₂ MW: 612.25 g/mol Melting Pt: 178…183 °C Storage Temperature: Ambient |
MDL Number:
MFCD12545948 CAS Number: 1002345-50-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...